Dibafit™ Barbed Adapters with 1/4"-28 UNF(F) Connection

Use adapters to quickly connect tubing lines or fittings
  • Easily connect soft tubing with a 1/4"-28 UNF(F) connection
Designed for use with soft wall tubing, these barbed adapters have a superior barb with a crisp edge to hold the tubing securely in place. Select from acetal or PEEK material of construction.
$120.67 - $137.47CAD / Pkg of 5

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (mm) Fitting Material
Availability Pricing
Dibafit™ Adapter, 1/4"-28 UNF(F) flat bottom to 1.0 mm ID, PEEK™; 5/pk 2194114 Mfr # 008NB10-KD2L
COMPARE
1 PEEK
$137.47CAD / Pkg of 5
Dibafit™ Adapter, 1/4"-28 UNF(F) flat bottom to 1.5 mm ID, PEEK™; 5/pk 2194118 Mfr # 008NB15-KD2L
COMPARE
1.5 PEEK
$137.47CAD / Pkg of 5
Dibafit™ Adapter, 1/4"-28 UNF(F) flat bottom to 2.0 mm ID, PEEK™; 5/pk 2194130 Mfr # 008NB20-KD2L
COMPARE
2 PEEK
$137.47CAD / Pkg of 5
Dibafit™ Adapter, 1/4"-28 UNF(F) flat bottom to 2.5 mm ID, PEEK™; 5/pk 2194134 Mfr # 008NB25-KD2L
COMPARE
2.5 PEEK
$120.67CAD / Pkg of 5
Dibafit™ Adapter, 1/4"-28 UNF(F) flat bottom to 3.2 mm ID, PEEK™; 5/pk 2194148 Mfr # 008NB32-KD2L
COMPARE
3.2 PEEK
$137.47CAD / Pkg of 5
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat