Diba MicroBarb Adapters, 1/4"-28 UNF(F)
Both the ID and OD of tubing are firmly gripped for a secure connection
  • Low-carryover connection for flexible to semi-rigid tubing
  • Chemically inert PEEK™ construction
Use MicroBarb adapters to securely connect flexible and semi-rigid tubing. Adapters feature two components—barbed component to fit inside the tube ID and threaded outer connector that slips over the tubing OD to secure the barb in place—to create a female threaded flat-bottom port.
$467.80 - $536.31CAD / Pkg of 25
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (mm) Compression Size / Tube Od (Port 1) (mm)
Availability Pricing
Diba MicroBarb® Adapter, 1/4"-28 UNF(F) to 0.01" ID x 0.081" OD, PEEK™; 25/pk 2194069 Mfr # 008MB025-208K2L
COMPARE
0.25 2.08
$536.31CAD / Pkg of 25
Diba MicroBarb® Adapter, 1/4"-28 UNF(F) to 0.01" ID x 0.092" OD, PEEK™; 25/pk 2194071 Mfr # 008MB025-234K2L
COMPARE
0.25 2.34
$467.80CAD / Pkg of 25
Diba MicroBarb® Adapter, 1/4"-28 UNF(F) to 0.02" ID x 0.092" OD, PEEK™; 25/pk 2194084 Mfr # 008MB051-234K2L
COMPARE
0.51 2.34
$467.80CAD / Pkg of 25
Diba MicroBarb® Adapter, 1/4"-28 UNF(F) to 0.02" ID x 1.043" OD, PEEK™; 25/pk 2194086 Mfr # 008MB051-264K2L
COMPARE
0.51 2.64
$467.80CAD / Pkg of 25
Diba MicroBarb® Adapter, 1/4"-28 UNF(F) to 0.03" ID x 0.082" OD, PEEK™; 25/pk 2194091 Mfr # 008MB076-208K2L
COMPARE
0.76 2.08
$536.31CAD / Pkg of 12
Diba MicroBarb® Adapter, 1/4"-28 UNF(F) to 0.037" ID x 0.103" OD, PEEK™; 25/pk 2194096 Mfr # 008MB095-263K2L
COMPARE
0.95 2.63
$536.31CAD / Pkg of 25