Continuous-Feed Grinding Drives
  • Continuous feed allows unlimited sample sizes
  • Two grinding heads available for one drive—interchange heads for different applications
  • Adjustable speed control from 3000 to 6,500 rpm
  • Continuous duty—great for inline applications
  • Surfaces that contact your samples are 304 SS—ideal for sanitary applications
  • Easy-to-clean stainless steel tray provides an ideal working surface
  • 1-1/3 hp (990-watt) motor is completely sealed from the grinding chamber by a dust-tight seal
Constructed of heavy-duty cast aluminum, enameled finish, with a stainless steel tray. Order grinding heads separately.
$7,910.63CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
IKA 2836001 Continuous feed grinding drive, 115 VAC/60 Hz 0430130 Mfr # 2836001
COMPARE
115
$7,910.63CAD / Each
IKA 2836000 Continuous feed grinding drive, 230 VAC/50 Hz 0430135 Mfr # 2836000
COMPARE
230
$7,910.63CAD / Each