Combination Filter/Ultraviolet Sterilizers
Filtration and sterilization in one compact unit!
  • Filters particulates down to 0.5 µm; UV system kills 99.9% of bacteria
These filter/sterilizers feature a UV bulb in the center of a carbon filter cartridge—the filter system removes chemicals and sediment, while the UV system kills 99.9% of the bacteria. UV bulb life is 9000 hours. Sterilizers feature an extended pass carbon block (EPCB) filter designed to provide longer contact with the UV system. Filter has 0.5 µm pore size to effectively remove chemicals, sediment, and Giardia cysts.
Order an optional faucet kit, replacement filter, and replacement UV bulb separately.
$817.73 - $1,395.30CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Combination Filter/Ultraviolet Sterilizer, 1 gpm, 120 VAC 9928000 Mfr # 15910111
COMPARE
Combination Filter/Ultraviolet Sterilizer, 1 gpm, 120 VAC
$817.73CAD / Each
Combination Filter/Ultraviolet Sterilizer; 2 GPM, 120 VAC 9928012 Mfr # 15610111
COMPARE
Combination Filter/Ultraviolet Sterilizer; 2 GPM, 120 VAC
$1,395.30CAD / Each
Combination Filter/Ultraviolet Sterilizer; 2 GPM, 220 VAC 9928017 Mfr # 15610211 + 42905002
COMPARE
Combination Filter/Ultraviolet Sterilizer; 2 GPM, 220 VAC
$1,298.79CAD / Each