Cole-Parmer® Zahn-Style Viscosity Cups
Simple and convenient – no complicated training needed
  • Quick measurement of the viscosity of liquids
  • Constructed of one-piece, corrosion resistant ASTM (899-94) series 400 stainless steel
  • Suspension handle holding cup
Measure the viscosity of liquids such as paint, lacquer, oil, and syrups. Simply insert the cup into your sample and lift the viscosity cup out, starting a stopwatch/timer. Cup is made of series 400 stainless steel and requires 44 mL of sample for the test. After withdrawing the cup, measure the time it takes until the first break in the liquid stream. A conversion sheet is supplied that will convert the Zahn time to a centistokes value.
The suspension handle holding cup is 12.078 in (30.51 cm) for dipping into liquids.
$285.47 - $311.77CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Cup Number Nist-Traceable Viscosity
Availability Pricing
COMPARE
1 No 18 to 56 centistokes
$311.77CAD / Each
COMPARE
2 No 40 to 230 centistokes
$285.47CAD / Each
COMPARE
3 No 150 to 790 centistokes
$304.77CAD / Each
COMPARE
4 No 220 to 1100 centistokes
$309.85CAD / Each
COMPARE
5 No 460 to 1725 centistokes
$311.77CAD / Each