Cole-Parmer Viscosity Cup #5, 460 to 1725 Centistokes
Representative image only.
Cole-Parmer Viscosity Cup #5, 460 to 1725 Centistokes
Cole-Parmer | Item: 0870064
Simple and convenient – no complicated training needed
$311.77CAD / Each
$311.77CAD / Each
Buy It Together
Current Item
Cole-Parmer Viscosity Cup #5, 460 to 1725 centistokes
Cole-Parmer Viscosity Cup #5, 460 to 1725 centistokes
0870064
$311.77CAD / Each
+ + + =
Total Price 2380.12 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Instructions and conversion charts.
Key Features
  • Quick measurement of the viscosity of liquids
  • Constructed of one-piece, corrosion resistant ASTM (899-94) series 400 stainless steel
  • Suspension handle holding cup
  • Hole outside diameter is 0.208?
  • Range is 45-80 sec
More About this Item
Measure the viscosity of liquids such as paint, lacquer, oil, and syrups. Simply insert the cup into your sample and lift the viscosity cup out, starting a stopwatch/timer. Cup is made of series 400 stainless steel and requires 44 mL of sample for the test. After withdrawing the cup, measure the time it takes until the first break in the liquid stream. A conversion sheet is supplied that will convert the Zahn time to a centistokes value.
The suspension handle holding cup is 12.078 in (30.51 cm) for dipping into liquids.
Specifications
  • Viscosity460 to 1725 centistokes
  • Cup Number5
  • Nist-TraceableNo
  • MaterialStainless steel
  • DescriptionViscosity Cup #5, 460 to 1725 centistokes