Cole-Parmer® Utility Tanks with Covers, SS
Work on gas, electric, or even induction style stoves
  • Cylindrical tank with heat-resistant handles — cover included
  • Unsealed, medium gauge 304 SS construction suitable for induction-type heating
  • Encapsulated-aluminum bottom with three layers for even heat distribution and less energy consumption
Utility tanks (stock pots) are dishwasher safe and the brushed finish makes them easy to clean — perfect for use on gas, electric, and induction style stoves.
$453.07 - $1,411.51CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Utility Tank with Lid, 304 Stainless Steel, 11 L; Each
$453.07CAD / Each
COMPARE
Utility Tank with Lid, 304 Stainless Steel, 17 L; Each
$512.74CAD / Each
COMPARE
Utility Tank with Lid, 304 Stainless Steel, 26 L; Each
$682.92CAD / Each
COMPARE
Utility Tank with Lid, 304 Stainless Steel, 50 L; Each
$1,031.37CAD / Each
COMPARE
Utility Tank with Lid, 304 Stainless Steel, 71 L; Each
$1,411.51CAD / Each