Cole-Parmer Utility Tank with Lid, 304 Stainless Steel, 11 L; Each
Representative image only.
Cole-Parmer Utility Tank with Lid, 304 Stainless Steel, 11 L; Each
Cole-Parmer | Item: 0723103
Work on gas, electric, or even induction style stoves
$453.07CAD / Each
BUY MORE & SAVE
  • 1-5$453.07 CAD / Each
  • 6-29$407.76 CAD / Each
  • 30-119$385.11 CAD / Each
  • 120+$362.46 CAD / Each
$453.07CAD / Each
BUY MORE & SAVE
  • 1-5$453.07 CAD / Each
  • 6-29$407.76 CAD / Each
  • 30-119$385.11 CAD / Each
  • 120+$362.46 CAD / Each
Product Description
Key Features
  • Cylindrical tank with heat-resistant handles — cover included
  • Unsealed, medium gauge 304 SS construction suitable for induction-type heating
  • Encapsulated-aluminum bottom with three layers for even heat distribution and less energy consumption
  • 1 year warranty
More About this Item
Utility tanks (stock pots) are dishwasher safe and the brushed finish makes them easy to clean — perfect for use on gas, electric, and induction style stoves.
1 Year Warranty
Specifications
  • MaterialMedium-gauge 304 stainless steel with encapsulated-aluminum bottom (three layers)
  • Volume (Gal)2.9
  • Volume (Liters)11
  • Width (in)9 7/16
  • Width (cm)24
  • Height (in)9 7/16
  • Height (cm)24
  • DescriptionUtility Tank with Lid, 304 Stainless Steel, 11 L; Each