Cole-Parmer® TrayCell Cuvette and Caps

The TrayCell is a fibre optic cuvette which can be used with as little as 0.7 µl of sample
  • Ideal for DNA, RNA and protein measurements
  • Sample concentration range from 25 to 4250 µg/ml
  • Sample is simply wiped away after the measurements, making the TrayCell quick and easy to use
  • Prevents dilution and pipetting errors
  • Fits all standard 10 x 10 mm cuvette holders and micro-cuvette holders
The cuvette is supplied with two caps which give path lengths of either 1 mm or 0.2 mm, thus creating a "virtual dilution" of 1:10 or 1:50 of the sample when compared to a measurement with a standard 10 mm cuvette. Additional or replacement caps can also be purchased separately.

Also for applications of very low or very high DNA concentrations, we can offer two addition caps to provide path lengths of 2 mm or 0.1 mm creating dilutions of 1:5 or 1:100.
$1,664.93 - $10,807.32CAD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Replacement 1 mm Path Length Cap for TrayCell
$1,937.51CAD / Each
COMPARE
Replacement 0.2 mm Path Length Cap for TrayCell
$1,937.51CAD / Each
COMPARE
Additional 2 mm Path Length Cap for TrayCell
$1,664.93CAD / Each
COMPARE
Additional 0.1 mm Path Length Cap for TrayCell
$1,664.93CAD / Each
COMPARE
TrayCell Cuvette for Ultra-Micro Sample Volumes
$10,807.32CAD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat