Cole-Parmer 035 263 Replacement 1 mm Path Length Cap for TrayCell

Representative image only.

Cole-Parmer 035 263 Replacement 1 mm Path Length Cap for TrayCell

Cole-Parmer | Item: 7900048

The TrayCell is a fibre optic cuvette which can be used with as little as 0.7 µl of sample

$1,937.51CAD / Each

$1,937.51CAD / Each
Product Description

Key Features

  • Ideal for DNA, RNA and protein measurements
  • Sample concentration range from 25 to 4250 µg/ml
  • Sample is simply wiped away after the measurements, making the TrayCell quick and easy to use
  • Prevents dilution and pipetting errors
  • Fits all standard 10 x 10 mm cuvette holders and micro-cuvette holders

More About this Item

The cuvette is supplied with two caps which give path lengths of either 1 mm or 0.2 mm, thus creating a "virtual dilution" of 1:10 or 1:50 of the sample when compared to a measurement with a standard 10 mm cuvette. Additional or replacement caps can also be purchased separately.

Also for applications of very low or very high DNA concentrations, we can offer two addition caps to provide path lengths of 2 mm or 0.1 mm creating dilutions of 1:5 or 1:100.

Specifications
  • Pathlength (mm)1
  • DescriptionReplacement 1 mm Path Length Cap for TrayCell
Reviews
Q&A