Cole-Parmer® Testing Sieves, 8" dia, Brass Frame/Brass Wire
Get more accurate results
  • Sievers are serialized
  • Come with a test sieve certificate of compliance
These brass testing sieves are serialized and come with a test sieve certificate of compliance. Equivalent to ASTM E11-09 compliance sieve.
$152.13 - $433.55CAD / Each
48 variations of this product are available.
See All Product Options Below
48 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (InchShort) Sieve Opening (mm) U.S. Standard Screen Size (No.)
Availability Pricing
COMPARE
2 2.36 8
$157.65CAD / Each
COMPARE
2 2 10
$157.65CAD / Each
COMPARE
2 1.68 12
$157.65CAD / Each
COMPARE
2 1.41 14
$157.65CAD / Each
COMPARE
2 1.19 16
$157.65CAD / Each
COMPARE
2 1 18
$157.65CAD / Each
COMPARE
2 0.841 20
$157.65CAD / Each
COMPARE
2 0.707 25
$214.75CAD / Each
COMPARE
2 0.595 30
$157.65CAD / Each
COMPARE
2 0.5 35
$157.65CAD / Each
COMPARE
2 0.425 40
$157.65CAD / Each
COMPARE
2 0.355 45
$157.65CAD / Each
COMPARE
2 0.297 50
$157.65CAD / Each
COMPARE
2 0.25 60
$214.75CAD / Each
COMPARE
2 0.212 70
$157.65CAD / Each
COMPARE
2 0.177 80
$157.65CAD / Each
COMPARE
2 0.149 100
$157.65CAD / Each
COMPARE
2 0.125 120
$153.23CAD / Each
COMPARE
2 0.105 140
$166.86CAD / Each
COMPARE
2 0.074 200
$204.80CAD / Each
Showing 1 - 20 of 48