Cole-Parmer Sterile Gridded Cellulose Nitrate Membranes
White colour with black grid facilitates colony counting
Economical cellulose nitrate (CN) membranes come sterilized by EtO and are individually wrapped. These hydrophilic membranes do not contain any extractable to inhibit or stimulate growth of bacteria, do not affect pH of the medium, and are suitable for both aqueous and protein filtration. Membranes may be exposed to temperatures up to 55°C (131°F). Additional variations are available; call us for details.
$113.01 - $113.16CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Pore Size (µm)
Availability Pricing
COMPARE
47 Cellulose Nitrate 0.2
$113.16CAD / Each
COMPARE
47 Cellulose Nitrate 0.45
$113.01CAD / Each