Cole-Parmer® Stainless Steel Tweezers with Carbon Fiber Tips
Great for handling electronic components safely
  • ESD-safe tips combined with rugged stainless steel handles
  • Carbon fiber tip offers excellent abrasion and wear resistance.
Polyamide reinforced with 30% carbon fiber construction provides highly rigid tip with great tensile and flexural strength. Stainless steel handle offers excellent corrosion resistance to most chemicals, salts, and acids. These tweezers are ideal for handling sensitive components and devices such as in the electronic industry. Antimagnetic and anti-acid.
$77.79CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
ESD Safe Overall Length (in) Description
Availability Pricing
COMPARE
Yes 5 Stainless Steel Tweezers w/ Flat, Thick, Plastic Tips
$77.79CAD / Each
COMPARE
Yes 5 Stainless Steel Tweezers w/ Flat, Round, Plastic Tips
$77.79CAD / Each
COMPARE
Yes 5 Stainless Steel Tweezers w/ Very Sharp, Tapered, Fine, Plastic Tips
$77.79CAD / Each
COMPARE
Yes 5 Stainless Steel Tweezers w/ Sharp, Pointed, Plastic Tips
$77.79CAD / Each