Cole-Parmer 00CF.SA.1 Stainless Steel Tweezers w/ Flat, Thick, Plastic Tips
Representative image only.
Cole-Parmer 00CF.SA.1 Stainless Steel Tweezers w/ Flat, Thick, Plastic Tips
Cole-Parmer | Item: 0738727
Great for handling electronic components safely
$77.79CAD / Each
$77.79CAD / Each
Product Description
Key Features
  • ESD-safe tips combined with rugged stainless steel handles
  • Carbon fiber tip offers excellent abrasion and wear resistance.
  • Limited Lifetime Warranty
More About this Item
Polyamide reinforced with 30% carbon fiber construction provides highly rigid tip with great tensile and flexural strength. Stainless steel handle offers excellent corrosion resistance to most chemicals, salts, and acids. These tweezers are ideal for handling sensitive components and devices such as in the electronic industry. Antimagnetic and anti-acid.
Limited Lifetime Warranty
Specifications
  • Overall Length (in)5
  • Overall Length (cm)12.5
  • MaterialCarbon Fiber-Reinforced Polyamide-Tipped Stainless Steel
  • SterileNo
  • ESD SafeYes
  • DescriptionStainless Steel Tweezers w/ Flat, Thick, Plastic Tips
  • WarrantyLimited Lifetime