Cole-Parmer® Specific Gravity Plain Form Precision Hydrometers for Light Liquids
  • Temperature of Standardization 60°F/60°F
  • Metal Ballast
  • Individually Serialized
Each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory. Individually packaged with a multi-language Statement of Accuracy.
$60.41 - $89.43CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Precision 0.760/0.830 Specific Gravity Hydrometer
$60.41CAD / Each
COMPARE
Precision 0.820/0.890 Specific Gravity Hydrometer
$84.13CAD / Each
COMPARE
Precision 0.880/0.950 Specific Gravity Hydrometer
$89.43CAD / Each
COMPARE
Precision 0.940/1.010 Specific Gravity Hydrometer
$84.57CAD / Each