Cole-Parmer Specific Gravity Plain Form Hydrometers for Light Liquids
  • Statement of accuracy
  • Temperature of Standardization 60°F/60°F
  • Metal Ballast
  • Individually Serialized
Each hydrometer is manufactured in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
$46.41 - $53.93CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
0.690/0.800 Specific Gravity Hydrometer for Liquids Lighter Than Water
$46.41CAD / Each
COMPARE
0.790/0.900 Specific Gravity Hydrometer for Liquids Lighter Than Water
$46.56CAD / Each
COMPARE
0.890/1.000 Specific Gravity Hydrometer for Liquids Lighter Than Water
$46.41CAD / Each
COMPARE
0.940/1.010 Specific Gravity Hydrometer for Liquids Lighter Than Water
$46.41CAD / Each
COMPARE
0.700/0.810 Specific Gravity Hydrometer for Liquids Lighter Than Water
$49.65CAD / Each
COMPARE
0.800/0.910 Specific Gravity Hydrometer for Liquids Lighter Than Water
$48.62CAD / Each
COMPARE
0.900/1.000 Specific Gravity Hydrometer for Liquids Lighter Than Water
$48.62CAD / Each
COMPARE
0.600/0.670 Specific Gravity Hydrometer for Liquids Lighter Than Water
$53.93CAD / Each
Showing 1- 8 of 8