Cole-Parmer® Specific Gravity & Baume Dual Scale Hydrometers for Liguids Lighter than Water
  • NIST Traceable
  • Temperature of Standardization 60°F/60°F
  • Metal Ballast
  • Individually Serialized
For liquids lighter than water, each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
$68.81 - $76.32CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
0.700/1.000 Specific Gravity and 10/70 Degree Baume Dual Scale Hydrometer for Liquids Lighter Than Water
$68.81CAD / Each
COMPARE
0.790/0.900 Specific Gravity and 24/45 Degree Baume Dual Scale Hydrometer for Liquids Lighter Than Water
$72.20CAD / Each
COMPARE
0.890/1.000 Specific Gravity and 10/25 Degree Baume Dual Scale Hydrometer for Liquids Lighter Than Water
$76.32CAD / Each