Cole-Parmer® Single Glass Cuvettes for Visible Spectroscopy
Transmission is over 80% at 365nm
  • Standard-size cuvettes are the type most often used for routine laboratory testing
  • Cuvettes include chemical resistant, PTFE covers
  • Cuvettes have two clear sides and two frosted sides
These Cole-Parmer® single glass cuvettes are constructed of glass and suitable for use in visible spectroscopy applications.
$113.16 - $163.55CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Max Capacity (mL) Pathlength (mm)
Availability Pricing
COMPARE
Glass 0.4 1
$115.37CAD / Each
COMPARE
Glass 0.7 2
$125.39CAD / Each
COMPARE
Glass 1.7 5
$113.16CAD / Each
COMPARE
Glass 3.5 10
$116.25CAD / Each
COMPARE
Glass 7 20
$123.91CAD / Each
COMPARE
Glass 10.5 30
$130.84CAD / Each
COMPARE
Glass 14 40
$132.75CAD / Each
COMPARE
Glass 17.5 50
$163.55CAD / Each
Showing 1- 8 of 8