Cole-Parmer® Polystat® Digital Plus Stainless Steel Open-Top Bath
On-screen prompts make digital controller easy to set up and operate
  • Highly-visible, swiveling controller displays and controls bath temperatures with ±0.01°C precision
  • Touchpad control accompanies 3-3/4” screen
  • Automatic performance optimization and specific heat tuning enhance precision
  • On-screen prompts and message bar simplify operation
Well-suited for a wide variety of laboratory applications, the sleek stainless steel bath is accompanied by a Digital Plus controller allowing you to customize operation through a variety of control settings.
Set pump speed, specific heat value, calibration offset and internal or external temperature control in a straightforward, intuitive fashion. The swiveling controller also features a message bar that assists in making changes and provides alerts and safety information.
User safety is enhanced by redundant over temperature protection, low-liquid level float switch and settable high and low temperature limits.
The controller supports four languages (English, Spanish, French and German) while bidirectional RS-232, addressable RS-485 and Ethernet communications facilitate remote control and data logging. USB ports enable you to log data directly to a flash drive or computer.
$4,412.80 - $5,229.06CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Reservoir Capacity (L)
Availability Pricing
COMPARE
150 120 6
$4,412.80CAD / Each
COMPARE
150 240 6
$4,412.80CAD / Each
COMPARE
150 120 10
$4,530.67CAD / Each
COMPARE
150 120 20
$4,781.15CAD / Each
COMPARE
150 240 20
$4,781.15CAD / Each
COMPARE
150 120 28
$5,229.06CAD / Each
COMPARE
150 240 28
$5,229.06CAD / Each
Showing 1- 7 of 7