Cole-Parmer® Polar Endcapped C18 HPLC & UHPLC Columns and Accessories
Specially designed for separation and retention of polar analytes
  • Retention of polars by polar endcapping group
  • Excellent reproducibility and robustness
  • Fully scalable
Reduce silanol activity and protect the silica from dissolution by endcapping the HPLC columns. Endcapped columns are 100% aqueous compatible and excellent for separating and retaining polar analytes.
Columns come in 316L stainless steel (SS) tubing hardware. A SS female end fitting with a SS and PEEK frit system keeps the stationary phase in place and packed bed from disturbance. The SS end fitting accepts a male 10-32 threaded nut with standard 1/16" OD tubing.
Use the guard cartridges to maintain chromatographic integrity. They ensure erroneous materials are kept out of the more important and expensive analytical columns, leading to more accurate results. Simply match the sizes to select the correct guard cartridge for your column; guard cartridge holder 16470-89 is required to use the guard cartridges. In-line filters are available for your UPHLC and HPLC columns—order separately.
$581.99 - $1,140.40CAD / Each
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Particle Size (µm) Description
Availability Pricing
COMPARE
4.6 5 Polar Endcapped C18 HPLC Column, 5 µm, 250 mm L x 4.6 mm ID; 1/EA
$1,029.90CAD / Each
COMPARE
4.6 5 Polar Endcapped C18 HPLC Column, 5 µm, 150 mm L x 4.6 mm ID; 1/EA
$994.54CAD / Each
COMPARE
4.6 5 Polar Endcapped C18 HPLC Column, 5 µm, 100 mm L x 4.6 mm ID; 1/EA
$975.38CAD / Each
COMPARE
2.1 5 Polar Endcapped C18 HPLC Column, 5 µm, 50 mm L x 2.1 mm ID; 1/EA
$882.56CAD / Each
COMPARE
4.6 3 Polar Endcapped C18 HPLC Column, 3 µm, 150 mm L x 4.6 mm ID; 1/EA
$1,075.57CAD / Each
COMPARE
4.6 3 Polar Endcapped C18 HPLC Column, 3 µm, 100 mm L x 4.6 mm ID; 1/EA
$1,057.89CAD / Each
COMPARE
2.1 3 Polar Endcapped C18 HPLC Column, 3 µm, 150 mm L x 2.1 mm ID; 1/EA
$1,001.91CAD / Each
COMPARE
2.1 3 Polar Endcapped C18 HPLC Column, 3 µm, 100 mm L x 2.1 mm ID; 1/EA
$984.22CAD / Each
COMPARE
2.1 3 Polar Endcapped C18 HPLC Column, 3 µm, 50 mm L x 2.1 mm ID; 1/EA
$975.38CAD / Each
COMPARE
4.6 1.7 Polar Endcapped C18 UHPLC Column, 1.7 µm, 50 mm L x 4.6 mm ID; 1/EA
$1,140.40CAD / Each
COMPARE
2.1 1.7 Polar Endcapped C18 UHPLC Column, 1.7 µm, 150 mm L x 2.1 mm ID; 1/EA
$1,082.94CAD / Each
COMPARE
2.1 1.7 Polar Endcapped C18 UHPLC Column, 1.7 µm, 100 mm L x 2.1 mm ID; 1/EA
$1,066.73CAD / Each
COMPARE
2.1 1.7 Polar Endcapped C18 UHPLC Column, 1.7 µm, 50 mm L x 2.1 mm ID; 1/EA
$1,049.05CAD / Each
COMPARE
4.6 5 Polar Endcapped C18 Guard Cartridges, 5 µm, 10 mm L x 4.6 mm ID; 2/PK
$581.99CAD / Pkg of 2
COMPARE
4.6 5 Polar Endcapped C18 Guard Cartridges, 5 µm, 10 mm L x 4.6 mm ID; 4/PK
$1,044.63CAD / Pkg of 4
COMPARE
2.1 5 Polar Endcapped C18 Guard Cartridges, 5 µm, 10 mm L x 2.1 mm ID; 2/PK
$581.99CAD / Pkg of 2
COMPARE
2.1 5 Polar Endcapped C18 Guard Cartridges, 5 µm, 10 mm L x 2.1 mm ID; 4/PK
$1,044.63CAD / Pkg of 4
COMPARE
4.6 3 Polar Endcapped C18 Guard Cartridges, 3 µm, 10 mm L x 4.6 mm ID; 2/PK
$581.99CAD / Pkg of 2
COMPARE
4.6 3 Polar Endcapped C18 Guard Cartridges, 3 µm, 10 mm L x 4.6 mm ID; 4/PK
$1,044.63CAD / Pkg of 4
COMPARE
2.1 3 Polar Endcapped C18 Guard Cartridges, 3 µm, 10 mm L x 2.1 mm ID; 2/PK
$581.99CAD / Pkg of 2
Showing 1 - 20 of 21