Cole-Parmer® Plus Class A "TD" Graduated Cylinders
Heavy hexagonal base provides stability to prevent cylinder from tipping over
  • Class A, Type 1 borosilicate glass withstands temperatures from -58°F to 1022°F (-50°C to 550°C)
  • Graduations are durable white enamel
  • Autoclavable
Reference: ASTM E1272 Class A tolerances and specifications.
$76.76 - $171.28CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
COMPARE
50 1 Borosilicate Glass
$76.76CAD / Each
COMPARE
100 1 Borosilicate Glass
$92.09CAD / Each
COMPARE
250 2 Borosilicate Glass
$165.39CAD / Each
COMPARE
500 5 Borosilicate Glass
$171.28CAD / Each