Cole-Parmer® Pathogen Contamination Detection Kits for qPCR
Quickly and accurately verify pathogen contamination in a food source
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Great value at 100 tests per kit
Use the speciation kits to clarify sources and types of products where accurate results are critical. Order a complete kit with mastermix or select a kit without mastermix and use your own.
$1,550.01 - $2,277.86CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Binomial Name Mastermix Included Test Type
Availability Pricing
COMPARE
Legionella pneumophila No DNA
$1,550.01CAD / Each
COMPARE
JC Virus No DNA
$1,819.64CAD / Each
COMPARE
JC Virus Yes DNA
$2,277.86CAD / Each
COMPARE
Bacteria domain No DNA
$1,819.64CAD / Each
COMPARE
Bacteria domain Yes DNA
$2,277.86CAD / Each
COMPARE
Clostridium estertheticum No DNA
$1,819.64CAD / Each
COMPARE
Mycobacterium avium subspecies paratuberculosis No DNA
$1,550.01CAD / Each
COMPARE
Mycobacterium avium subspecies paratuberculosis Yes DNA
$1,938.98CAD / Each
Showing 1- 8 of 8