Cole-Parmer DNA JC Virus, With Mastermix
Representative image only.
Cole-Parmer DNA JC Virus, With Mastermix
Cole-Parmer | Item: 9394401
Quickly and accurately verify pathogen contamination in a food source
$2,277.86CAD / Each
$2,277.86CAD / Each
Product Description
Includes
Lyophilized mastermix, lyophilized ROX, resuspension buffer, lyophilized primer and probe mix, positive control for standard curve, internal extraction control DNA, endogenous control, and RNAse/DNAse-free water.
Key Features
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Great value at 100 tests per kit
More About this Item
Use the speciation kits to clarify sources and types of products where accurate results are critical. Order a complete kit with mastermix or select a kit without mastermix and use your own.
Specifications
  • Test TypeDNA
  • Number of Tests Included150
  • Mastermix IncludedYes
  • Binomial NameJC Virus
  • DescriptionJC Virus, With Mastermix