Cole-Parmer MSS-200 Trinocular Stereozoom Microscope
Robust and sturdy microscope that provides crisp, high-resolution images
  • 0.7x to 4.5x zoom
  • Trinocular 45° inclined tubes with 360° rotatable head
  • Incident and transmitted adjustable LED illuminations
  • Coarse adjustment with tension control
  • Choose the stand that works best for your needs
Cole-Parmer trinocular microscopes feature paired wide field eyepieces WF10x/20 mm with eyecups. The head features an adjustable diopter with an interpupillary distance adjustable between 55 and 75 mm. The incident and transmitted adjustable LED illuminations can be used at the same time. Microscope is constructed of an alloy metal cast and is stove hardened off-white painted with a transparent and black/white stage plant.
$1,171.35 - $1,215.55CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Stand Type Description
Availability Pricing
COMPARE
100 to 240 Pillar Trinocular Stereozoom Microscope, Pillar Stand, 7.0-45x zoom, 100-240 VAC
$1,171.35CAD / Each
COMPARE
100 to 240 Rack and pinion Trinocular Stereozoom Microscope, Rack and Pinion Stand, 7.0-45x zoom, 100-240 VAC
$1,215.55CAD / Each