Cole-Parmer® Mini Serological Pipettes
An alternative for cramped working spaces, this mini pipette saves you 10.2 cm (4") off standard pipettes!
  • Easy-to-read graduations from +1 - 2%
  • Color-coded rings for easy identification
  • Polystyrene(PS), gamma sterilized and pyrogen free
  • Unique filter plug stays put and eliminates lint fibers
Developed for use in cramped spaces such as laminar hoods, these mini serological pipettes measure 22.4 cm (9") long and fit most serological pipettors. The shorter length reduces distance between sample and pipettor, aiding in more ergonomic pipetting. Gamma sterilized; individually-wrapped with easy peel-apart opening. Dispenser box lets you quickly access pipettes without opening the box top.
$113.45 - $183.44CAD / Case of 200
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Sterile Description
Availability Pricing
COMPARE
5000 Yes Mini Serological Pipette, Polystyrene, 5 mL, Blue; 200/cs
$154.34CAD / Case of 200
COMPARE
10000 Yes Mini Serological Pipette, Polystyrene, 10 mL, Orange; 150/cs
$113.45CAD / Case of 150
COMPARE
25000 Yes Mini Serological Pipette, Polystyrene, 25 mL, Green; 100/cs
$183.44CAD / Case of 100