Cole-Parmer® Low-Temperature PCR Racks
Safely store PCR samples down to -80 °C
  • Uniquely designed lid also works as an elevated rack base
  • Easily stack racks to save benchtop and freezer space
  • Alphanumeric identification provides fast location and retrieval of tubes
  • Each rack holds 96 x 0.2 mL tubes
Securely store 0.2 mL PCR tubes in these durable ABS plastic low-temperature racks. Racks feature textured finger grips for easier handling and transportation. Alphanumeric identification offers fast sample and well identification. Simply flip the translucent polycarbonate lid to use it as an elevated rack base. Side openings on the lids provide faster cooling ensuring the integrity of the samples.
$41.84CAD / Pkg of 5
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Tubes Held Description
Availability Pricing
COMPARE
Assorted 96 Low-Temperature PCR Rack, 96-Well, Assorted; 5/PK
$41.84CAD / Pkg of 5
COMPARE
Blue 96 Low-Temperature PCR Rack, 96-Well, Blue; 5/PK
$41.84CAD / Pkg of 5
COMPARE
Green 96 Low-Temperature PCR Rack, 96-Well, Green; 5/PK
$41.84CAD / Pkg of 5