Cole-Parmer® Low-Form Graduated Glass Cylinders
Accurate measurements even at lowest graduations
  • Calibrated To Deliver
  • Double metric scale
Two sets of graduations let you read these cylinders up or down. Cylinders provide accurate measurements even at the lowest graduations. Low-profile design and wider base reduce chances of accidental tipping. Cylinders feature easy-to-read white scales and two pouring spouts.
$14.73 - $16.65CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
COMPARE
50 1 Glass
$14.73CAD / Each
COMPARE
100 2 Glass
$16.65CAD / Each