Cole-Parmer® Lab Sampling Scoops, Stainless Steel
Durable and strong, fully welded handle
  • Stainless steel construction for high chemical resistance
  • Rounded scoop with high-quality, mirror-smooth finish allows for easy and complete cleaning
  • Reusable; provides a more sustainable option to disposable scoops
These seamless, stainless steel lab scoops are for general purpose use and perfect for chemicals and foods. The angled, fully welded handle provides for easy, comfortable scooping and optimal durability in rugged applications. These sampling scoops have a mirror-smooth finish for thorough cleaning to help prevent cross contamination. They are autoclavable if sterilization is needed.
When used in place of disposable scoops, these reusable lab scoops help reduce waste in the laboratory, as well as save you money and reordering time. Indicated capacities are approximate and are not intended for measuring specific volumes.
$44.79 - $407.02CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Length (cm) Width (cm)
Availability Pricing
COMPARE
100 20.5 6.5
$44.79CAD / Each
COMPARE
200 22.5 8.5
$58.20CAD / Each
COMPARE
300 27.5 9
$72.64CAD / Each
COMPARE
500 29 10.5
$109.33CAD / Each
COMPARE
1000 30.2 12.5
$173.12CAD / Each
COMPARE
1183 30.5 19.1
$407.02CAD / Each