Cole-Parmer® Glass Cuvettes
Get accurate spectroscopic readings
  • Constructed of durable glass
Glass cuvettes are available in a wide capacity range from 500 µL to 35 mL and pathlengths from 10 to 100 mm. The glass cuvettes are suitable for wavelength range measurements of visible light.
Visible wavelength % transmission: maximum from 400 to 750 nm
$167.97 - $664.50CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Capacity (mL) Pathlength (mm) Description
Availability Pricing
COMPARE
7 20 Glass Cuvette, Visible, Up to 7 mL, 20 mm Path; 1/EA
$328.20CAD / Each
COMPARE
35 100 Glass Cuvette, Visible, Up to 35 mL, 100 mm Path; 1/EA
$664.50CAD / Each
COMPARE
0.5 10 Glass Cuvette, Visible, Micro, 500 uL, 10 mm Path; 1/EA
$404.08CAD / Each
COMPARE
1 10 Glass Cuvette, Visible, 1 mL, 10 mm Path; 1/EA
$525.26CAD / Each
COMPARE
1 10 Glass Cuvette, Visible, Black 1 mL, 10 mm Path; 1/EA
$436.49CAD / Each
COMPARE
3.5 10 Glass Cuvette, Visible, 2.0 to 3.5 mL, 10 mm Path; 1/EA
$167.97CAD / Each
COMPARE
14 40 Glass Cuvette, Visible, Up to 14 mL 40 mm Path; 1/EA
$352.51CAD / Each
COMPARE
17.5 50 Glass Cuvette, Visible, Up to 17.5 mL, 50 mm Path; 1/EA
$411.08CAD / Each
Showing 1- 8 of 8