Cole-Parmer® General Purpose Freestanding Undercounter/Countertop Freezers
Dependable freezing solutions for general purpose applications
  • Manual defrost system allows users to maintain optimal performance with periodic defrosting
  • Adjustable operating temperature from -15°C to -25°C
  • Built-In Security with a keyed door lock and magnetic door gasket to ensure a positive seal
  • Uses natural hydrocarbon refrigerants and meets UL/C-UL standards
Cole-Parmer® General Purpose Freestanding Undercounter/Countertop Freezers provide a practical and efficient solution for general storage needs where precise temperature control and advanced monitoring are not required. These units feature a manual defrost system to help maintain consistent performance, with an adjustable temperature range from -15°C to -25°C. Built-in security features, including a keyed door lock and a magnetic door gasket, ensure safe storage of your items. Designed with eco-friendly hydrocarbon refrigerants, these freezers offer a sustainable choice for laboratories and general-purpose settings.
ITEM(S) ON SALE
$1,032.56 - $1,667.88CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Freezer Capacity (cu ft) Number of Doors Power (VAC)
Availability Pricing
COMPARE
1.5 1 120
$1,032.56CAD / Each
$1,290.69CAD / Each
COMPARE
4 1 120
$1,667.88CAD / Each
$2,084.85CAD / Each