Cole-Parmer® Amino HPLC & UHPLC Columns and Accessories
Specifically designed for amino acid analysis
  • Rugged and reproducible amino bonding
  • Excellent for carbohydrate species
  • Rapid equilibration
The amino HPLC Columns feature highly pure, porous spherical silica gel for amino acid analysis—excellent for separation of sugars. Use these columns for reversed phase, normal phase, or ion-exchange separations.
Columns come in 316L stainless steel (SS) tubing hardware. A SS female end fitting with a SS and PEEK frit system keeps the stationary phase in place and packed bed from disturbance. The SS end fitting accepts a male 10-32 threaded nut with standard 1/16" OD tubing.
Use the guard cartridges to maintain chromatographic integrity. They ensure erroneous materials are kept out of the more important and expensive analytical columns, leading to more accurate results. Simply match the sizes to select the correct guard cartridge for your column; guard cartridge holder 16470-89 is required to use the guard cartridges. In-line filters are available for your UPHLC and HPLC columns—order separately.
$581.99 - $1,140.40CAD / Each
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Particle Size (µm) Description
Availability Pricing
COMPARE
4.6 5 Amino HPLC Column, 5 µm, 250 mm L x 4.6 mm ID; 1/EA
$1,029.90CAD / Each
COMPARE
4.6 5 Amino HPLC Column, 5 µm, 150 mm L x 4.6 mm ID; 1/EA
$994.54CAD / Each
COMPARE
4.6 5 Amino HPLC Column, 5 µm, 100 mm L x 4.6 mm ID; 1/EA
$975.38CAD / Each
COMPARE
2.1 5 Amino HPLC Column, 5 µm, 50 mm L x 2.1 mm ID; 1/EA
$882.56CAD / Each
COMPARE
4.6 3 Amino HPLC Column, 3 µm, 150 mm L x 4.6 mm ID; 1/EA
$1,085.89CAD / Each
COMPARE
4.6 3 Amino HPLC Column, 3 µm, 100 mm L x 4.6 mm ID; 1/EA
$1,066.73CAD / Each
COMPARE
2.1 3 Amino HPLC Column, 3 µm, 150 mm L x 2.1 mm ID; 1/EA
$1,012.22CAD / Each
COMPARE
2.1 3 Amino HPLC Column, 3 µm, 100 mm L x 2.1 mm ID; 1/EA
$994.54CAD / Each
COMPARE
2.1 3 Amino HPLC Column, 3 µm, 50 mm L x 2.1 mm ID; 1/EA
$975.38CAD / Each
COMPARE
4.6 1.7 Amino UHPLC Column, 1.7 µm, 50 mm L x 4.6 mm ID; 1/EA
$1,140.40CAD / Each
COMPARE
2.1 1.7 Amino UHPLC Column, 1.7 µm, 150 mm L x 2.1 mm ID; 1/EA
$1,082.94CAD / Each
COMPARE
2.1 1.7 Amino UHPLC Column, 1.7 µm, 100 mm L x 2.1 mm ID; 1/EA
$1,066.73CAD / Each
COMPARE
2.1 1.7 Amino UHPLC Column, 1.7 µm, 50 mm L x 2.1 mm ID; 1/EA
$1,049.05CAD / Each
COMPARE
4.6 5 Amino Guard Cartridges, 5 µm, 10 mm L x 4.6 mm ID; 2/PK
$581.99CAD / Pkg of 2
COMPARE
4.6 5 Amino Guard Cartridges, 5 µm, 10 mm L x 4.6 mm ID; 4/PK
$1,044.63CAD / Pkg of 4
COMPARE
2.1 5 Amino Guard Cartridges, 5 µm, 10 mm L x 2.1 mm ID; 2/PK
$581.99CAD / Pkg of 2
COMPARE
2.1 5 Amino Guard Cartridges, 5 µm, 10 mm L x 2.1 mm ID; 4/PK
$1,044.63CAD / Pkg of 4
COMPARE
4.6 3 Amino Guard Cartridges, 3 µm, 10 mm L x 4.6 mm ID; 2/PK
$581.99CAD / Pkg of 2
COMPARE
4.6 3 Amino Guard Cartridges, 3 µm, 10 mm L x 4.6 mm ID; 4/PK
$1,044.63CAD / Pkg of 4
COMPARE
2.1 3 Amino Guard Cartridges, 3 µm, 10 mm L x 2.1 mm ID; 2/PK
$581.99CAD / Pkg of 2
Showing 1 - 20 of 21