Cole-Parmer® Absorbent Benchtop Liner
Protect benchtop from spills and breakage
  • Prevents spills from spreading
  • Cushions surface while absorbing liquid
  • Soft and durable
  • Cut to fit depending on needs
Made of highly absorbent three-ply tissue with waterproof polypropylene backing that is resistant to most chemicals.
$108.59 - $499.48CAD / Roll of 1
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Width (in) Description
Availability Pricing
COMPARE
20 Absorbent Bench-Top Liner, 3 Ply with Poly Backing, 20" x 300', Roll
$267.42CAD / Roll of 1
COMPARE
20 Absorbent Bench-Top Liner, 3 Ply with Poly Backing, 20" x 300', 2 Roll/PK
$492.11CAD / Pkg of 2
COMPARE
20 Absorbent Bench-Top Liner, 3 Ply with Poly Backing, 20" x 50', 6 Roll/PK
$499.48CAD / Pkg of 6
COMPARE
20 Absorbent Bench-Top Liner, 3 Ply with Poly Backing, 20" x 50', Roll
$108.59CAD / Roll of 1
COMPARE
20 Absorbent Bench-Top Liner, 3 Ply, Poly Back, 20x24" Pre-Cut; 50/Pk
$185.28CAD / Pkg of 50