Cole-Parmer® 500- and 750-Watt Ultrasonic Processors with Temperature Controller
Process batch samples as small as 250 µL or as large as 1000 mL using optional continuous flow cell
  • Integral temperature controller and memory prevents harmful overheating of the sample by terminating the ultrasonics when the sample reaches a predetermined temperature limit
  • Exclusive energy (joules) monitor provides the most accurate method to reproduce results
  • On/off independent pulser helps prevent overheating
  • User-friendly menu-driven prompts provide intuitive guidance through all functions
These ultrasonic processors have the same features as the models above plus an integral temperature controller and memory. The temperature controller prevents harmful overheating of the sample by terminating the ultrasonics when the sample reaches a predetermined temperature limit (select from 1 to 100ºC); order temperature probe separately below. The time-saving memory stores up to ten procedures to facilitate protocol duplication, automate repetitive tasks, and eliminate user-to-user method variability for consistent reproducibility.
$9,223.42 - $9,886.45CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Watts
Availability Pricing
COMPARE
115 500
$9,223.42CAD / Each
COMPARE
230 500
$9,223.42CAD / Each
COMPARE
115 750
$9,886.45CAD / Each
COMPARE
230 750
$9,886.45CAD / Each