Coarse Sieve Shaker
Economical shaker produces consistent, accurate results
For use with materials coarser than 100 mesh (150 µm), this shaker includes quick-change adapter to accommodate either 8'' or 12'' diameter test sieves. The quiet shaking action provides consistent and effective results.
Benefit from its versatility, as shaker accommodates six full-height or thirteen half-height (8'' diameter) sieves; four full-height or six half-height (12'' diameter sieves). Oscillations per minute: 280 (60 Hz); 231 (50 Hz). Shpg wt 80 lb (36.3 kg).
$3,757.14 - $3,764.51CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
W.S. Tyler RX-812 Coarse Sieve Shaker, 8" or 12" sieves, 110 VAC 5998671 Mfr # RX-812
COMPARE
110
$3,757.14CAD / Each
W.S. Tyler RX-812-3 Coarse Sieve Shaker, 8" or 12" sieves, 220 VAC 5998678 Mfr # RX-812-3
COMPARE
220
$3,764.51CAD / Each