Chempette Digital Repetitive Pipettors
Ideal for prolonged pipetting applications
  • Effortless digital volume adjustment
  • Lockable for precision accuracy
  • Plastic housing protect glass cylinder and plunger
Dispense with ±1% full scale accuracy and ±0.2% full scale reproducibility. Easy to use—simply place TYGON® inlet tubing (20-5/8'' L, included) with PTFE sinker (included) into your vessel, then dial in desired digital volume setting. Volume setting lock prevents accidental reset. Borosilicate glass cylinder and plunger are enclosed in PP housing to prevent breakage.
Do not autoclave. Shpg wt 1 lb (0.5 kg).
$794.16 - $800.05CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy (%) Max Volume (mL) Min Volume (mL)
Availability Pricing
Chempette A3-21DG-1 Digital Repetitive Pipettor, 0.2 to 1 mL 0784810 Mfr # A3-21DG-1
COMPARE
±1.00% 1 0.2
$794.16CAD / Each
Chempette A3-21DG-5 Digital Repetitive Pipettor, 1 to 5 mL 0784830 Mfr # A3-21DG-5
COMPARE
±1.00% 5 1
$794.16CAD / Each
Chempette A3-21DG-10 Digital Repetitive Pipettor, 2 to 10 mL 0784840 Mfr # A3-21DG-10
COMPARE
±1.00% 10 2
$800.05CAD / Each