Cell Culture Spin Flasks
Perfect for small scale operations, or quick tests of your fermentation process
  • Designed for microcarrier and suspension cultures, such as insect cells, hybridomas, and adapted cell lines
  • Bottom dimple prevents cell crushing and improves circulation
Use the 45-mm sidearms as air vents, inoculation ports, or media inlet/outlet ports, or use sidearms to insert probes for pH, dissolved oxygen, or CO2 measurement and control. Flasks are made of borosilicate glass, and include a PTFE and glass impeller assembly. Sidearms have PTFE-faced silicone liners. Top (center) cap has two liners: one silicone and one PTFE (to be used together). Entire spin flask unit is autoclavable. Additional sizes are available; please call our Applications Specialists for details.
$976.12 - $1,116.83CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Description
Availability Pricing
DWK Life Sciences (Wheaton) 356882 Cell Culture Spin Flask, 500 mL; 1/Pk 2930000 Mfr # 356882
COMPARE
500 Glass Cell Culture Spin Flask, 500 mL; 1/Pk
$976.12CAD / Each
DWK Life Sciences (Wheaton) 356884 Cell Culture Spin Flask, 1-Liter; 1/Pk 2930002 Mfr # 356884
COMPARE
1000 Glass Cell Culture Spin Flask, 1-Liter; 1/Pk
$1,116.83CAD / Each