DWK Life Sciences (Wheaton) 356882 Cell Culture Spin Flask, 500 mL; 1/Pk

Representative image only.

DWK Life Sciences (Wheaton) 356882 Cell Culture Spin Flask, 500 mL; 1/Pk

DWK Life Sciences (Wheaton) | Mfr: 356882| Item: 2930000

Perfect for small scale operations, or quick tests of your fermentation process

$976.12CAD / Each

$976.12CAD / Each
Product Description

Includes

PTFE and glass impeller assembly.

Key Features

  • Designed for microcarrier and suspension cultures, such as insect cells, hybridomas, and adapted cell lines
  • Bottom dimple prevents cell crushing and improves circulation

More About this Item

Use the 45-mm sidearms as air vents, inoculation ports, or media inlet/outlet ports, or use sidearms to insert probes for pH, dissolved oxygen, or CO2 measurement and control. Flasks are made of borosilicate glass, and include a PTFE and glass impeller assembly. Sidearms have PTFE-faced silicone liners. Top (center) cap has two liners: one silicone and one PTFE (to be used together). Entire spin flask unit is autoclavable. Additional sizes are available; please call our Applications Specialists for details.

Specifications
  • MaterialGlass
  • Capacity (mL)500
  • Overall Height (cm)17
  • DescriptionCell Culture Spin Flask, 500 mL; 1/Pk
Reviews
Q&A