Burkle Disposable Polystyrene Food-Grade Sampling Scoops
EU and FDA compliant to safely handle food samples
  • Suitable for food-grade applications
  • Individually packaged in easy-to-open bags
  • Produced and assembled according to cleanroom ISO Class 7
These scoops are ideal for sampling powders, granulates, and pastes. A transparent cover, available on some scoops, safeguards against possible contamination and spillage. Specially designed handle allows scoops to stand horizontally, while an angled front edge prevents samples from falling out. The scoops come in seven size options and are available nonsterilized or presterilized via gamma irradiation.
$47.15 - $208.12CAD / Pkg of 10
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Sterile Description
Availability Pricing
Burkle 5378-0001 Disposable Sampling Scoop, PS, FDA Compliant, White; 25 mL 0604231 Mfr # 5378-0001
COMPARE
25 No Disposable Sampling Scoop, PS, FDA Compliant, White; 25 mL
$47.15CAD / Pkg of 10
Burkle 5378-0003 Disposable Sampling Scoop, PS, FDA Compliant, White; 50 mL 0604233 Mfr # 5378-0003
COMPARE
50 No Disposable Sampling Scoop, PS, FDA Compliant, White; 50 mL
$48.62CAD / Pkg of 10
Burkle 5378-0005 Disposable Sampling Scoop, PS, FDA Compliant, White; 100 mL 0604235 Mfr # 5378-0005
COMPARE
100 No Disposable Sampling Scoop, PS, FDA Compliant, White; 100 mL
$50.10CAD / Pkg of 10
Burkle 5378-0006 Disposable Sampling Scoop with Cover, PS, FDA Compliant, White; 100 mL 0604236 Mfr # 5378-0006
COMPARE
100 No Disposable Sampling Scoop with Cover, PS, FDA Compliant, White; 100 mL
$77.94CAD / Pkg of 10
Burkle 5378-0013 Disposable Sampling Scoop, PS, FDA Compliant, White; 250 mL 0604243 Mfr # 5378-0013
COMPARE
250 No Disposable Sampling Scoop, PS, FDA Compliant, White; 250 mL
$62.77CAD / Pkg of 10
Burkle 5378-0019 Disposable Sampling Scoop, PS, FDA Compliant, White; 1000 mL 0604247 Mfr # 5378-0019
COMPARE
1000 No Disposable Sampling Scoop, PS, FDA Compliant, White; 1000 mL
$143.21CAD / Pkg of 10
Burkle 5378-1001 Disposable Sampling Scoop, PS, FDA Compliant, White, Sterile; 25 mL 0604251 Mfr # 5378-1001
COMPARE
25 Yes Disposable Sampling Scoop, PS, FDA Compliant, White, Sterile; 25 mL
$51.72CAD / Pkg of 10
Burkle 5378-1002 Disposable Sampling Scoop with Cover, PS, FDA Compliant, White, Sterile; 25 mL 0604252 Mfr # 5378-1002
COMPARE
25 Yes Disposable Sampling Scoop with Cover, PS, FDA Compliant, White, Sterile; 25 mL
$78.68CAD / Pkg of 10
Burkle 5378-1003 Disposable Sampling Scoop, PS, 160 mm, FDA Compliant, White, Sterile; 50 mL 0604253 Mfr # 5378-1003
COMPARE
50 Yes Disposable Sampling Scoop, PS, 160 mm, FDA Compliant, White, Sterile; 50 mL
$53.93CAD / Pkg of 10
Burkle 5378-1004 Disposable Sampling Scoop with Cover, PS, FDA Compliant, White, Sterile; 50 mL 0604254 Mfr # 5378-1004
COMPARE
50 Yes Disposable Sampling Scoop with Cover, PS, FDA Compliant, White, Sterile; 50 mL
$80.89CAD / Pkg of 10
Burkle 5378-1005 Disposable Sampling Scoop, PS, FDA Compliant, White, Sterile; 100 mL 0604255 Mfr # 5378-1005
COMPARE
100 Yes Disposable Sampling Scoop, PS, FDA Compliant, White, Sterile; 100 mL
$55.99CAD / Pkg of 10
Burkle 5378-1007 Disposable Sampling Scoop, PS, FDA Compliant, White, Sterile; 150 mL 0604257 Mfr # 5378-1007
COMPARE
150 Yes Disposable Sampling Scoop, PS, FDA Compliant, White, Sterile; 150 mL
$57.02CAD / Pkg of 10
Burkle 5378-1013 Disposable Sampling Scoop, PS, FDA Compliant, White, Sterile; 250 mL 0604263 Mfr # 5378-1013
COMPARE
250 Yes Disposable Sampling Scoop, PS, FDA Compliant, White, Sterile; 250 mL
$69.54CAD / Pkg of 10
Burkle 5378-1014 Disposable Sampling Scoop with Cover, PS, FDA Compliant, White, Sterile; 250 mL 0604264 Mfr # 5378-1014
COMPARE
250 Yes Disposable Sampling Scoop with Cover, PS, FDA Compliant, White, Sterile; 250 mL
$117.72CAD / Pkg of 10
Burkle 5378-1015 Disposable Sampling Scoop, PS, FDA Compliant, White, Sterile; 500 mL 0604265 Mfr # 5378-1015
COMPARE
500 Yes Disposable Sampling Scoop, PS, FDA Compliant, White, Sterile; 500 mL
$122.29CAD / Pkg of 10
Burkle 5378-1016 Disposable Sampling Scoop with Cover, PS, FDA Compliant, White, Sterile; 500 mL 0604266 Mfr # 5378-1016
COMPARE
500 Yes Disposable Sampling Scoop with Cover, PS, FDA Compliant, White, Sterile; 500 mL
$208.12CAD / Pkg of 10
Burkle 5378-1019 Disposable Sampling Scoop, PS, FDA Compliant, White, Sterile; 1000 mL 0604267 Mfr # 5378-1019
COMPARE
1000 Yes Disposable Sampling Scoop, PS, FDA Compliant, White, Sterile; 1000 mL
$189.70CAD / Pkg of 10
Burkle 5378-3003 Disposable Sampling Scoop, PS, FDA Compliant, Blue, Sterile; 50 mL 0604272 Mfr # 5378-3003
COMPARE
50 Yes Disposable Sampling Scoop PS, FDA Compliant, Blue, Sterile; 50 mL
$53.93CAD / Pkg of 10
Burkle 5378-3007 Disposable Sampling Scoop, PS, FDA Compliant, Blue, Sterile; 150 mL 0604274 Mfr # 5378-3007
COMPARE
150 Yes Disposable Sampling Scoop PS, FDA Compliant, Blue, Sterile; 150 mL
$57.02CAD / Pkg of 10
Burkle 5378-3013 Disposable Sampling Scoop, PS, FDA Compliant, Blue, Sterile; 250 mL 0604278 Mfr # 5378-3013
COMPARE
250 Yes Disposable Sampling Scoop PS, FDA Compliant, Blue, Sterile; 250 mL
$69.54CAD / Pkg of 10
Showing 1- 20 of 20