Bud Industries NEMA 4X Galvanized and Stainless Steel Enclosures
Designed to meet NEMA Type 1, 2, 4, 4X, and IP65 ratings
  • Continuous seam welded 16-gauge steel or stainless steel construction
  • Hinged cover clamps to the base with keyed quarter-turn latch for a watertight seal
  • External mounting brackets so box can be mounted without opening cover
  • Bonding studs ground the base and door
  • Easily machined and modified for specific application
  • Smooth gray powder-coat finish; includes clear zinc plate internal panel and hardware
  • UL listed type 4, 12, and 13
$132.75 - $1,128.62CAD / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Outside Height (in) Outside Width (in)
Availability Pricing
Bud Industries SNB3730 NEMA 4X Galvanized Steel Enclosure 9450223 Mfr # SNB-3730
COMPARE
Steel 9.84 7.87
$152.13CAD / Each
Bud Industries SNB3731 NEMA 4X Galvanized Steel Enclosure 9450225 Mfr # SNB-3731
COMPARE
Steel 11.81 9.84
$229.11CAD / Each
Bud Industries SNB3732 NEMA 4X Galvanized Steel Enclosure 9450271 Mfr # SNB-3732
COMPARE
Steel 11.81 9.84
$241.64CAD / Each
Bud Industries SNB3734 NEMA 4X Galvanized Steel Enclosure 9450274 Mfr # SNB-3734
COMPARE
Steel 11.81 11.814
$280.31CAD / Each
Bud Industries SNB3736 NEMA 4X Galvanized Steel Enclosure 9450276 Mfr # SNB-3736
COMPARE
Steel 15.75 11.81
$243.11CAD / Each
Bud Industries SNB3737 NEMA 4X Galvanized Steel Enclosure 9450277 Mfr # SNB-3737
COMPARE
Steel 15.75 15.75
$337.41CAD / Each
Bud Industries SNB3738 NEMA 4X Galvanized Steel Enclosure 9450278 Mfr # SNB-3738
COMPARE
Steel 7.87 5.91
$132.75CAD / Each
Bud Industries SNB3741 NEMA 4X Galvanized Steel Enclosure 9450281 Mfr # SNB-3741
COMPARE
Steel 9.84 7.87
$159.49CAD / Each
Bud Industries SNB3743 NEMA 4X Galvanized Steel Enclosure 9450282 Mfr # SNB-3743
COMPARE
Steel 15.75 13.98
$402.24CAD / Each
Bud Industries SNB3731-SS NEMA 4X Stainless Steel Enclosure 9450284 Mfr # SNB-3731-SS
COMPARE
Stainless Steel 11.81 9.84
$702.81CAD / Each
Bud Industries SNB3732-SS NEMA 4X Stainless Steel Enclosure 9450285 Mfr # SNB-3732-SS
COMPARE
Stainless Steel 11.81 9.84
$662.29CAD / Each
Bud Industries SNB3733-SS NEMA 4X Stainless Steel Enclosure 9450286 Mfr # SNB-3733-SS
COMPARE
Stainless Steel 11.81 11.81
$839.83CAD / Each
Bud Industries SNB3734-SS NEMA 4X Stainless Steel Enclosure 9450287 Mfr # SNB-3734-SS
COMPARE
Stainless Steel 11.81 11.814
$857.51CAD / Each
Bud Industries SNB3736-SS NEMA 4X Stainless Steel Enclosure 9450289 Mfr # SNB-3736-SS
COMPARE
Stainless Steel 15.75 11.81
$1,022.53CAD / Each
Bud Industries SNB3738-SS NEMA 4X Stainless Steel Enclosure 9450291 Mfr # SNB-3738-SS
COMPARE
Stainless Steel 7.87 5.91
$407.02CAD / Each
Bud Industries SNB3739-SS NEMA 4X Stainless Steel Enclosure 9450292 Mfr # SNB-3739-SS
COMPARE
Stainless Steel 7.87 7.87
$286.21CAD / Each
Bud Industries SNB3740-SS NEMA 4X Stainless Steel Enclosure 9450293 Mfr # SNB-3740-SS
COMPARE
Stainless Steel 9.84 9.84
$591.57CAD / Each
Bud Industries SNB3741-SS NEMA 4X Stainless Steel Enclosure 9450294 Mfr # SNB-3741-SS
COMPARE
Stainless Steel 9.84 7.87
$405.55CAD / Each
Bud Industries SNB3743-SS NEMA 4X Stainless Steel Enclosure 9450295 Mfr # SNB-3743-SS
COMPARE
Stainless Steel 15.75 13.98
$1,128.62CAD / Each
Showing 1- 19 of 19