Bud Industries SNB3733-SS NEMA 4X Stainless Steel Enclosure
Representative image only.
Bud Industries SNB3733-SS NEMA 4X Stainless Steel Enclosure
Bud Industries | Mfr: SNB-3733-SS| Item: 9450286
Designed to meet NEMA Type 1, 2, 4, 4X, and IP65 ratings
$839.83CAD / Each
$839.83CAD / Each
Product Description
Includes
Internal panel and hardware.
Key Features
  • Continuous seam welded 16-gauge steel or stainless steel construction
  • Hinged cover clamps to the base with keyed quarter-turn latch for a watertight seal
  • External mounting brackets so box can be mounted without opening cover
  • Bonding studs ground the base and door
  • Easily machined and modified for specific application
  • Smooth gray powder-coat finish; includes clear zinc plate internal panel and hardware
  • UL listed type 4, 12, and 13
  • 1 year warranty
1 Year WarrantyIP 65
Specifications
  • MaterialStainless Steel
  • Outside Width (in)11.81
  • Outside Height (in)11.81
  • Outside Depth (in)5.91
  • DescriptionNEMA 4X Stainless Steel Enclosure