BrandTech® Polypropylene Test Tube Racks
Durable polypropylene racks available in a variety of sizes and colors
  • Choice of racks that hold 21, 32, 40, 55 or 84 test tubes with various diameters
  • Polypropylene racks snap together for easy assembly
  • Stackable, non-conductive with alphanumerical markings for easy identification
  • Will not float in water baths
These sturdy BRAND® test tube racks keep their stability when submerged in water. The multipurpose racks are great in water baths, as well as transporting or storing test tubes. The racks hold test tubes with the various diameters: up to 30 mm using the 21-tube (7 x 3) rack; up to 25 mm for the 32-tube (8 x 4) rack, and up to 20 mm for the 40-tube (4 x 10) rack. The two versions of the 55-tube (5 x 11) racks can hold tubes with diameters up to 16 mm and 18 mm, while the 84-tube (6 x 14) rack, up to 13 mm in diameter.
They have an operating temperature range of -20 to 90°C, and autoclavable at 121°C. The racks are 265 mm long and 88 mm tall for the 21-, 32- and 40-tube racks, and 265 mm long and 75 mm tall for the 55- and 84-tube racks; all in which have a width of about 126 mm.
$163.91CAD / Pkg of 5
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Max Tube Size (mm) Tubes Held
Availability Pricing
BrandTech 4340000 Test Tube Rack, Polypropylene, White, 84 Tubes up to 13 mm dia.; 5/PK 0610100 Mfr # 4340000
COMPARE
White 13 84
$163.91CAD / Pkg of 5
BrandTech 4340001 Test Tube Rack, Polypropylene, Blue, 84 Tubes up to 13 mm dia.; 5/PK 0610101 Mfr # 4340001
COMPARE
Blue 13 84
$163.91CAD / Pkg of 5
BrandTech 4340010 Test Tube Rack, Polypropylene, White, 55 Tubes up to 18 mm Dia.; 5/PK 0610103 Mfr # 4340010
COMPARE
White 18 55
$163.91CAD / Pkg of 5
BrandTech 4340011 Test Tube Rack, Polypropylene, Blue, 55 Tubes up to 18 mm dia.; 5/PK 0610104 Mfr # 4340011
COMPARE
Blue 18 55
$163.91CAD / Pkg of 5
BrandTech 4340012 Test Tube Rack, Polypropylene, Red, 55 Tubes up to 18 mm dia.; 5/PK 0610105 Mfr # 4340012
COMPARE
Red 18 55
$163.91CAD / Pkg of 5
BrandTech 4340020 Test Tube Rack, Polypropylene, White, 40 Tubes up to 20 mm dia.; 5/PK 0610107 Mfr # 4340020
COMPARE
White 20 40
$163.91CAD / Pkg of 5
BrandTech 4340040 Test Tube Rack, Polypropylene, White, 21 Tubes up to 30 mm dia.; 5/PK 0610114 Mfr # 4340040
COMPARE
White 30 21
$163.91CAD / Pkg of 5
BrandTech 4340041 Test Tube Rack, Polypropylene, Blue, 21 Tubes up to 30 mm dia.; 5/PK 0610115 Mfr # 4340041
COMPARE
Blue 30 21
$163.91CAD / Pkg of 5
BrandTech 4340042 Test Tube Rack, Polypropylene, Red, 21 Tubes up to 30 mm dia.; 5/PK 0610116 Mfr # 4340042
COMPARE
Red 30 21
$163.91CAD / Pkg of 5
Showing 1- 9 of 9