BrandTech® Dispensette® S Organic Bottletop Dispensers
Best choice for concentrated acids and organic solvents
  • Proven floating piston design provides one handed operation even for large volumes
  • Numerous safety features
  • Chemically resistant materials, such as PTFE, ETFE, PFA, FEP, Al203, platinum-iridium and borosilicate glass
  • Hinged screw cap hinges away from dispensing when removed
  • Available with priming valve
This fourth-generation Dispensette® S Organic bottletop dispenser is an excellent choice for general dispensing. A reinforced discharge tube with swing-away screw cap improves safety while an improved fluid path requires even lower operating forces. A new safety discharge valve reduces the chance of dispensing unless the discharge tube is mounted properly.
Choose a dispenser with a recirculation valve if you want chemicals to route back into the reagent bottle during the priming step. Analog models feature adjustable volume selection with interior scalloped track to provide reproducibe settings.
Dispensette® S Organic Dispensers are optimized for use with concentrated acids and organic solvents. Not for use with hydrofluoric acid.
$1,012.22 - $2,350.06CAD / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Division (mL) Max Dispense Volume (mL) Min Dispense Volume (mL)
Availability Pricing
BrandTech Dispensette® S Organic Bottletop Dispenser, Fixed Volume, with Stantard Valve, 5 mL 0791547 Mfr # 4630230
COMPARE
5
$1,012.22CAD / Each
BrandTech Dispensette® S Organic Bottletop Dispenser, Fixed Volume, with Stantard Valve, 10 mL 0791548 Mfr # 4630240
COMPARE
10
$1,012.22CAD / Each
BrandTech Dispensette® S Organic Bottletop Dispenser, Digital Adjustable, with Stantard Valve, 0.5-5 mL 0791553 Mfr # 4630330
COMPARE
0.02 5 0.5
$1,174.29CAD / Each
BrandTech Dispensette® S Organic Bottletop Dispenser, Digital Adjustable, with Stantard Valve, 1-10 mL 0791554 Mfr # 4630340
COMPARE
0.05 10 1
$1,174.29CAD / Each
BrandTech Dispensette® S Organic Bottletop Dispenser, Digital Adjustable, with Stantard Valve, 2.5-25 mL 0791555 Mfr # 4630350
COMPARE
0.1 25 2.5
$1,583.89CAD / Each
BrandTech Dispensette® S Organic Bottletop Dispenser, Digital Adjustable, with Stantard Valve, 5-50 mL 0791556 Mfr # 4630360
COMPARE
0.2 50 5
$1,613.36CAD / Each
BrandTech Dispensette® S Organic Bottletop Dispenser, Fixed Volume, with Recirculation Valve, 10 mL 0791558 Mfr # 4630241
COMPARE
10
$1,077.05CAD / Each
BrandTech Dispensette® S Organic Bottletop Dispenser, Digital Adjustable, with Recirculation Valve, 0.5-5 mL 0791561 Mfr # 4630331
COMPARE
0.02 5 0.5
$1,237.65CAD / Each
BrandTech Dispensette® S Organic Bottletop Dispenser, Digital Adjustable, with Recirculation Valve, 1-10 mL 0791562 Mfr # 4630341
COMPARE
0.05 10 1
$1,237.65CAD / Each
BrandTech Dispensette® S Organic Bottletop Dispenser, Digital Adjustable, with Recirculation Valve, 2.5-25 mL 0791563 Mfr # 4630351
COMPARE
0.1 25 2.5
$1,650.20CAD / Each
BrandTech Dispensette® S Organic Bottletop Dispenser, Digital Adjustable, with Recirculation Valve, 5-50 mL 0791564 Mfr # 4630361
COMPARE
0.2 50 5
$1,679.66CAD / Each
BrandTech Dispensette® S Organic Bottletop Dispenser, Analog Adjustable, with Stantard Valve, 0.5-5 mL 0791576 Mfr # 4630130
COMPARE
0.1 5 0.5
$1,012.22CAD / Each
BrandTech Dispensette® S Organic Bottletop Dispenser, Analog Adjustable, with recirculation valve, 0.5-5 mL 0791577 Mfr # 4630131
COMPARE
0.1 5 0.5
$1,077.05CAD / Each
BrandTech Dispensette® S Organic Bottletop Dispenser, Analog Adjustable, with Stantard Valve, 1-10 mL 0791578 Mfr # 4630140
COMPARE
0.2 10 1
$1,012.22CAD / Each
BrandTech Dispensette® S Organic Bottletop Dispenser, Analog Adjustable, with Recirculation Valve, 1-10 mL 0791579 Mfr # 4630141
COMPARE
0.2 10 1
$1,077.05CAD / Each
BrandTech Dispensette® S Organic Bottletop Dispenser, Analog Adjustable, with Recirculation Valve, 2.5-25 mL 0791581 Mfr # 4630151
COMPARE
0.5 25 2.5
$1,480.76CAD / Each
BrandTech Dispensette® S Organic Bottletop Dispenser, Analog Adjustable, with Recirculation Valve, 5-50 mL 0791583 Mfr # 4630161
COMPARE
1 50 5
$1,517.59CAD / Each
BrandTech Dispensette® S Organic Bottletop Dispenser, Analog Adjustable, with Recirculation Valve, 10-100 mL 0791585 Mfr # 4630171
COMPARE
1 100 10
$2,350.06CAD / Each
Showing 1- 18 of 18