BrandTech Dispensette® S Organic Bottletop Dispenser, Analog Adjustable, with recirculation valve, 0.5-5 mL

Representative image only.

BrandTech Dispensette® S Organic Bottletop Dispenser, Analog Adjustable, with recirculation valve, 0.5-5 mL

BrandTech | Mfr: 4630131| Item: 0791577

Best choice for concentrated acids and organic solvents

$1,077.05CAD / Each

$1,077.05CAD / Each

Buy It Together

Current Item

BrandTech Dispensette® S Organic Bottletop Dispenser, Analog Adjustable, with recirculation valve, 0.5-5 mL
BrandTech Dispensette® S Organic Bottletop Dispenser, Analog Adjustable, with recirculation valve, 0.5-5 mL
0791577

$1,077.05CAD / Each
+ =
Total Price 1146.30 ADD 2 ITEMS TO CART
Product Description

Includes

45-mm standard thread plus polypropylene adapters: GL32-33, GL38, S40. Volumes 1, 2, 5, and 10 mL also include GL24-25 and GL28.

Key Features

  • Proven floating piston design provides one handed operation even for large volumes
  • Numerous safety features
  • Chemically resistant materials, such as PTFE, ETFE, PFA, FEP, Al203, platinum-iridium and borosilicate glass
  • Hinged screw cap hinges away from dispensing when removed
  • Available with priming valve
  • 1 year warranty

More About this Item

This fourth-generation Dispensette® S Organic bottletop dispenser is an excellent choice for general dispensing. A reinforced discharge tube with swing-away screw cap improves safety while an improved fluid path requires even lower operating forces. A new safety discharge valve reduces the chance of dispensing unless the discharge tube is mounted properly.
Choose a dispenser with a recirculation valve if you want chemicals to route back into the reagent bottle during the priming step. Analog models feature adjustable volume selection with interior scalloped track to provide reproducibe settings.
Dispensette® S Organic Dispensers are optimized for use with concentrated acids and organic solvents. Not for use with hydrofluoric acid.

1 Year Warranty
Specifications
  • Min Dispense Volume (mL)0.5
  • Max Dispense Volume (mL)5
  • Division (mL)0.1
  • Accuracy±0.5%
  • Reproducibility≤ 0.1%
  • AutoclavableYes
  • Wetted Partsborosilicate glass, aluminum oxide ceramic, tantalum, ETFE, FEP, PFA, PTFE, and PP
  • DescriptionOrganic Dispenser, 0.5 - 5 mL, analog, recirculating
  • Warranty1 year
Reviews
Q&A