Boekel Jitterbug Microplate Incubator Shaker
Accommodates two standard microplates or strip plates
  • Temperature, shaking speed, and mixing duration can be set independently.
  • Inner lid covers samples — reduces risk of biohazard contamination.
  • Double chamber design provides excellent temperature uniformity.
  • Ideal for a variety of immunoassay and research procedures
What's included: a power pack transformer with an 8-ft power cord and plug (three-prong plug on 115 VAC model; standard European plug on 230 VAC model).
$5,819.89 - $6,070.37CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Boekel Scientific 130000 Jitterbug Microplate Incubator Shaker, 115 VAC, 50/60 Hz 1305900 Mfr # 130000
COMPARE
115
$5,819.89CAD / Each
Boekel Scientific 130000-2 Jitterbug Microplate Incubator Shaker 230 VAC, 50/60 Hz 1305905 Mfr # 130000-2
COMPARE
230
$6,070.37CAD / Each