Boekel Scientific 130000 Jitterbug Microplate Incubator Shaker, 115 VAC, 50/60 Hz

Representative image only.

Boekel Scientific 130000 Jitterbug Microplate Incubator Shaker, 115 VAC, 50/60 Hz

Boekel Scientific | Mfr: 130000| Item: 1305900

Accommodates two standard microplates or strip plates

$5,819.89CAD / Each

$5,819.89CAD / Each
Product Description

Key Features

  • Temperature, shaking speed, and mixing duration can be set independently.
  • Inner lid covers samples — reduces risk of biohazard contamination.
  • Double chamber design provides excellent temperature uniformity.
  • Ideal for a variety of immunoassay and research procedures
  • 2 year warranty

More About this Item

What's included: a power pack transformer with an 8-ft power cord and plug (three-prong plug on 115 VAC model; standard European plug on 230 VAC model).

UL Listed to Canadian safety standards.Two year warranty
Specifications
  • Orbit Size (in)0.04
  • Orbit Size (mm)1
  • Max Speed (rpm)1500
  • Width (in)10
  • Height (in)10
  • Depth (in)4
  • Width (cm)25.4
  • Height (cm)25.4
  • Depth (cm)10.16
  • Max Operating Temperature (° F)77
  • Max Operating Temperature (° C)40
  • Temperature Uniformity±0.4°C with cover
  • TimerYes
  • Timer Range1 second to 99 minutes 59 seconds
  • DisplayLED
  • Power (VAC)115
  • Power (Hz)50/60
  • DescriptionJitterbug Microplate Incubator Shaker, 115 VAC, 50/60 Hz
  • Warranty2 year
Reviews
Q&A