Autoclavable Tape
Know your labware is autoclaved
  • Dark-colored diagonal mark revealed after autoclaving at 250°F (121°C) for 15 minutes
  • Sticks to glass, cloth, or metal
  • Leaves no residue
Crepe tape displays a dark colored diagonal after being autoclaved at 250°F (121°C) for 15 minutes. Adhesive backing sticks to glass, cloth or metal. Tape peels from surfaces easily and leaves no residue. Autoclavable
$31.53CAD / Roll of 1
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Material Description
Availability Pricing
COMPARE
41.67 Crepe Autoclave Tape with Indicating Label; 1" W; 500 in/RL, 1 RL/Pk
$31.53CAD / Roll of 1