Atago® Brix Refractometers
Precision accuracy ensures process and quality control across the entire Brix range!
  • Easy calibration via screw potentiometer
  • Automatic temperature compensation (ATC) models ideal for samples with temperature fluctuations
  • Made of polybutylene terephthalate (PBT)
Use these Brix refractometers to measure concentration of fruits, beverages, and canned goods; total solid content of syrups, vinegar, and sauces; or check the quality of vegetable, fruit, and grass tissues. Refractometers can also be used for chemical, agricultural, manufacturing process control, quality control, preshipment, and other applications.
$709.73 - $875.64CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Brix Range (%) Resolution
Availability Pricing
Atago MASTER-ALPHA Handheld Refractometer; 0.0 to 32.0% Brix, ATC, IP65 0294185 Mfr # 2311 MASTER-ALPHA
COMPARE
±0.2% (Brix) 32 0.2% (Brix)
$875.64CAD / Each
Atago 2312 MASTER-53T Handheld Refractometer; 0.0 to 53% Brix, ATC 0294187 Mfr # 2352 MASTER-53T
COMPARE
±0.2% (Brix) 53 0.2% (Brix)
$709.73CAD / Each