Ansell TouchNTuff® 92-575 Powdered Disposable Nitrile Glove
Keep hands dry and protected from chemical splashes
  • Textured fingertips provide secure grip
  • Complies with FDA food handling requirements
  • Ambidextrous
These robust nitrile gloves provide added protection for food processing and heavy-duty jobs. Powdered for easier donning and moisture absorption, the gloves are ideal for transferring liquids and solids, processing samples, handling incoming good, maintenance, and applications that have food contact.
$40.52 - $376.16CAD / Box of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Material Size
Availability Pricing
Ansell TouchNTuff® 92-575 Powdered Disposable Nitrile Gloves, Size M (7, 5-8); 100/Box 8161041 Mfr # 105126 BOX
COMPARE
9 1/2 Nitrile Medium
$40.52CAD / Box of 100
Ansell TouchNTuff® 92-575 Powdered Disposable Nitrile Gloves, Size L (8, 5-9); 100/Box 8161042 Mfr # 105127 BOX
COMPARE
9 1/2 Nitrile Large
$43.32CAD / Box of 100
Ansell TouchNTuff® 92-575 Powdered Disposable Nitrile Gloves, Size L (8, 5-9); 1000/CS (10 boxes of 100 gloves) 8162063 Mfr # 105127
COMPARE
9 1/2 Nitrile Large
$376.16CAD / Case of 1000
Ansell TouchNTuff® 92-575 Powdered Disposable Nitrile Gloves, Size XL (9, 5-10); 1000/CS (10 boxes of 100 gloves) 8162064 Mfr # 105128
COMPARE
9 1/2 Nitrile X-Large
$372.62CAD / Case of 1000