Ansell TouchNTuff® 92-575 Powdered Disposable Nitrile Gloves, Size XL (9, 5-10); 1000/CS (10 boxes of 100 gloves)
Representative image only.
Ansell TouchNTuff® 92-575 Powdered Disposable Nitrile Gloves, Size XL (9, 5-10); 1000/CS (10 boxes of 100 gloves)
Ansell | Mfr: 105128| Item: 8162064
Keep hands dry and protected from chemical splashes
$372.62CAD / Case of 1000
$372.62CAD / Case of 1000
Product Description
Key Features
  • Textured fingertips provide secure grip
  • Complies with FDA food handling requirements
  • Ambidextrous
More About this Item
These robust nitrile gloves provide added protection for food processing and heavy-duty jobs. Powdered for easier donning and moisture absorption, the gloves are ideal for transferring liquids and solids, processing samples, handling incoming good, maintenance, and applications that have food contact.
Specifications
  • Thickness (mil)5.5
  • MaterialNitrile
  • SizeX-Large
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlue
  • GripTextured fingertips
  • CuffStandard
  • ComplianceFDA 21 CFR 177-2600
  • SterileNo
  • PowderedYes
  • DescriptionPowdered Disposable Nitrile Gloves, Size XL (9,5-10)