Alicat Whisper™ MWB-Series Low-Pressure Drop Mass Flowmeters, Digital Display, Battery Operated, SCCM
Provides accurate real-time mass and volumetric flow readings with minimal pressure drop
  • Instant warm-up
  • Ninety-eight pre-loaded gas calibrations
  • One-hundred millisecond control response
  • Built-in rechargeable battery with up to 18 hours life
  • IP40 rating
These portable, low-pressure drop mass flowmeters utilize laminar differential pressure technology to deliver precise mass flow measurements for over 130 different gases. With a rapid control response time of just 100 milliseconds, they integrate easily into test stands, enabling immediate and accurate verification without the need for warm-up.
Designed to minimize pressure drop—as low as 0.07 psid—these flowmeters are ideal for applications with limited source pressure. Each unit comes pre-loaded with 98 standard gas calibrations that can be easily switched without compromising accuracy.
These stand-alone devices measure mass flow, volumetric flow, temperature, and absolute pressure, with readings shown in standard cubic centimeters per minute (SCCM). Additional features include a backlit monochrome LCD display, a data I/O port for seamless data logging to a computer, and versatile mounting options—even allowing for upside-down installation.
$2,902.58 - $3,735.04CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (SCCM) Accuracy Repeatability
Availability Pricing
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter, 0 - 0.5 SCCM, Battery Operated 6780315 Mfr # MWB-0.5SCCM-D
COMPARE
0.5 ±0.8% of reading ±0.2% of full scale ±0.2% of reading ±0.02% of full scale
$3,735.04CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter,  0 - 1 SCCM, Battery Operated 6780316 Mfr # MWB-1SCCM-D
COMPARE
1 ±0.8% of reading ±0.2% of full scale ±0.2% of reading ±0.02% of full scale
$3,735.04CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter,  0 - 2 SCCM, Battery Operated 6780317 Mfr # MWB-2SCCM-D
COMPARE
2 ±0.8% of reading ±0.2% of full scale ±0.2% of reading ±0.02% of full scale
$3,735.04CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter,  0 - 5 SCCM, Battery Operated 6780318 Mfr # MWB-5SCCM-D
COMPARE
5 ±0.8% of reading ±0.2% of full scale ±0.2% of reading ±0.02% of full scale
$2,902.58CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter,  0 - 10 SCCM, Battery Operated 6780319 Mfr # MWB-10SCCM-D
COMPARE
10 ±0.75% of reading ±0.1% of full scale ±0.1% of reading ±0.02% of full scale
$2,902.58CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter,  0 - 20 SCCM, Battery Operated 6780320 Mfr # MWB-20SCCM-D
COMPARE
20 ±0.75% of reading ±0.1% of full scale ±0.1% of reading ±0.02% of full scale
$2,902.58CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter,  0 - 50 SCCM, Battery Operated 6780321 Mfr # MWB-50SCCM-D
COMPARE
50 ±0.75% of reading ±0.1% of full scale ±0.1% of reading ±0.02% of full scale
$2,902.58CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter,  0 - 100 SCCM, Battery Operated 6780322 Mfr # MWB-100SCCM-D
COMPARE
100 ±0.75% of reading ±0.1% of full scale ±0.1% of reading ±0.02% of full scale
$2,902.58CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter, 0 - 200 SCCM, Battery Operated 6780323 Mfr # MWB-200SCCM-D
COMPARE
200 ±0.75% of reading ±0.1% of full scale ±0.1% of reading ±0.02% of full scale
$2,902.58CAD / Each
Alicat Whisper™ Low-Pressure Drop Mass Flowmeter, 0 - 500 SCCM, Battery Operated 6780324 Mfr # MWB-500SCCM-D
COMPARE
500 ±0.75% of reading ±0.1% of full scale ±0.1% of reading ±0.02% of full scale
$2,902.58CAD / Each
Showing 1- 10 of 10